MMP inhibitor profiling kit
Lieferant: ENZO LIFE SCIENCES AG
BMLAK3080001
ENZOBMLAK3080001EA
1090.1
EUR
ENZOBMLAK3080001
MMP inhibitor profiling kit
Assays
Enzymtests
The MMP inhibitor profiling kit, fluorometric RED is a complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes, using a quenched fluorogenic substrate OMNIMMP® RED.
The assays are performed in a convenient 96-well microplate format. The compound NNGH is also included as a prototypic control inhibitor.