Cathepsin S drug discovery kit
Lieferant: ENZO LIFE SCIENCES AG
BMLAK4310001
ENZOBMLAK4310001EA
589.6
EUR
ENZOBMLAK4310001
Cathepsin S drug discovery kit
Assays
Enzymtests
A QuantiZyme™ Assay System. The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit.
- Complete system designed to screen modulators of cathepsin S activity
- The assays are performed in a convenient 96-well microplate format
Lieferumfang: Kit contains Cathepsin S enzyme (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer and ½-volume white microplate.