Luminol (3-Aminophthalhydrazid) ≥95% (durch HPLC), CELLESTIAL®, ultrarein

Lieferant: ENZO LIFE SCIENCES AG
Achtung

Synonyme: NSC 5064

ENZ52354
ENZOENZ52354EA 136.4 EUR
ENZOENZ52354
Luminol (3-Aminophthalhydrazid) ≥95% (durch HPLC), CELLESTIAL®, ultrarein
Luminol (3-Aminophthalhydrazid)

ROS dye


Luminol is a luminescent peroxidase substrate that can be used as a chemiluminescent reagent for the determination of viable mammalian cells and bacteria. Luminol also has forensic application as a presumptive test for latent blood detection and is commonly employed in detection strategies by flow injection analysis, for pharmaceutical and environmental characterization. Chemiluminescent and electrochemiluminescent probes based upon luminal are beginning to merge with biochip and microarray development as a possible substitutes for the more wellestablished fluorescent probes.
Wavelength Maxima: Excitation 355nm, Emission 411nm

Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00006890
CAS-Nummer: 521-31-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e41862ab05ed5a3fe77d5cbe476e27a1f9eea5f&vid=fc15bac062192d863f9025f3d9240e62506dff2b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR