Thermal cyclers, Mastercycler® pro, individual modules
Lieferant: EPPENDORF
6320000.007
EPPE6320000.007EA 0 EUR
EPPE6320000.007
Thermal cyclers, Mastercycler® pro, individual modules
PCR-Cycler
Mastercycler pro is an open and flexible system which offers flexibility with fast heating and cooling rates. Systems are available as complete packages or separate modules.
  • Temperature control range 4 to 99 °C
  • Operates in three different configurations
  • Optional self-test dongle available (please enquire for details)
* A Control Panel or CycleManager pro software (both sold separately) is required for operation. CAN-Bus connection cables are required to link cyclers in a network.