C8- Aggresive cleanroom wiper size 12x12 150/bag, 18 bags/case. It is made of polycellulose material enchanced with aggressive pattern of hills/valley for pickup of enhanced contamination particles 1 * 150 ST
Lieferant: ESSENTRA POROUS TECHNOLOGIES
C8- Aggresive cleanroom wiper size 12x12 150/bag, 18 bags/case. It is made of polycellulose material enchanced with aggressive pattern of hills/valley for pickup of enhanced contamination particles 1 * 150 ST