MaxDetail® Maginifying Spectacles (for near) - Dark Grey
Lieferant: ESCHENBACH OPTIK
1624511
ESCH1624511EA 114 EUR
ESCH1624511
MaxDetail® Maginifying Spectacles (for near) - Dark Grey
Lupen
These hands-free, head-mounted glasses provide 2X magnification and can be adjusted so that each eye lens can be focused separately [±3 diopters], making the system adaptable to most patients. The glasses will focus on near objects with working distances of 25,4 mm and are perfect for any near viewing activity where a magnified image is helpful.
  • Lens Type: Galilean
  • Lens Material: PXM Plastic
  • Frame Size: 137 mm
  • Temple Size: 135 mm
  • Lens Size: 1,25
  • Field of view: 20°
  • PD Range: 60 to 80 mm
Bridge Size: Open saddle
Weight: 0,04 kg
Miscellaneous: Can be adjusted (+/- 3D) for spherical correction. The 1624-511 features dark grey temples and replaces the 1624-51 which had lighter colored temples.