Binocular, magno®
Lieferant: ESCHENBACH OPTIK
84177821
ESCH84177821EA 58.2 EUR
ESCH84177821
Binocular, magno®
Ferngläser und Monokulare
Suitable for a nature getaway, holiday or city break.
  • Small-sized, light and handy
  • Compact striking design for every occasion
  • Multi-layered coating ensuring sharp and true-colour images
Weight: 180 g
Bestellinformation: Includes a case and carrying cord.