DpnI, FastDigest™, fermentas
Lieferant: Thermo Fisher Scientific
FD1704
FD1704EA 70.75 EUR
FD1704
DpnI, FastDigest™, fermentas
Nukleinsäure-Reagenzien Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in oneuniversal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications