Digoxin

Lieferant: Fitzgerald Industries

10-D05-M91286
FITZ10-D05-M91286EA 0 EUR
FITZ10-D05-M91286
Digoxin
Digoxin
Formel: C₄₁H₆₄O₁₄
Molecular Weight: 780,95 g/mol
MDL: MFCD00003674
CAS-Nummer: 20830-75-5
EINECS: 244-068-1
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR