Serotonin

Lieferant: Fitzgerald Industries

10-S06-M09203
FITZ10-S06-M09203EA 0 EUR
FITZ10-S06-M09203
Serotonin
Serotonin
Formel: C₁₀H₁₂N₂O
CAS-Nummer: 50-67-9
EINECS: 200-058-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR