Hydrocortison

Lieferant: Fitzgerald Industries

Synonyme: Cortisol, 11β,17α,21-Trihydroxy-4-pregnen-3,20-dion, 11β,17α,21-Trihydroxypregn-4-en-3,20-dion, 11β-Hydroxycortison

20-CG50
FITZ20-CG50EA 0 EUR
FITZ20-CG50
Hydrocortison
Hydrocortison
Formel: C₂₁H₃₀O₅
Molecular Weight: 362,47 g/mol
Siedepunkt: 567 °C (1013 hPa)
Schmelzpunkt: 202…204 °C
Dichte: 1,4 g/cm³ (20 °C)
MDL: MFCD00011654
CAS-Nummer: 50-23-7
EINECS: 200-020-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR