Folsäure

Lieferant: Fitzgerald Industries

Synonyme: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

30-AF19
FITZ30-AF19EA 0 EUR
FITZ30-AF19
Folsäure
Folsäure
Formel: C₁₉H₁₉N₇O₆
Molecular Weight: 441,41 g/mol
MDL: MFCD00079305
CAS-Nummer: 59-30-3
EINECS: 200-419-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR