WR-Vespel+Edelstahl Greifgerät,M800-200S Manual wand (Tweezers), sized for correct handling of 150 mm (6 inch) and 200 mm (8 inch) wafers. The tweezers are of stainless steel with Vespel contact surfaces. Vespel is suitable for use up to 288 Deg C. T 1 * 1 Pack.
Lieferant: WINDRUSH TECHNOLOGY LTD.