5-Cyan-1-pentin
Lieferant: FLUOROCHEM
Synonyme:
5-Hexinnitril, 5-Hexinsäurenitril
009109-5G
FLUO009109-5GEA
222
EUR
FLUO009109-5G
5-Cyan-1-pentin
5-Cyan-1-pentin
Formel:
C₆H₇N Molecular Weight: 93,13 g/mol Siedepunkt: 115 °C (1013 hPa) Dichte: 0,891 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001978 CAS-Nummer: 14918-21-9 UN: 1992 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...