3-Formyl-6-isopropylchromon

Lieferant: CHEMPUR

FL12008.POR 012008-5G
CHMPFL12008.POREA 0 EUR
CHMPFL12008.POR FLUO012008-5G
3-Formyl-6-isopropylchromon
3-Formyl-6-isopropylchromon
Formel: C₁₃H₁₂O₃
Molecular Weight: 216,2326 g/mol
MDL: MFCD00192183
CAS-Nummer: 49619-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR