6-Fluor-3-formylchromon

Lieferant: CHEMPUR

Synonyme: 6-Fluorochromone-3-carboxaldehyde, 6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

FL12009.POR 012009-5G
CHMPFL12009.POREA 0 EUR
CHMPFL12009.POR FLUO012009-5G
6-Fluor-3-formylchromon
6-Fluor-3-formylchromon
Formel: C₁₀H₅FO₃
Molecular Weight: 192,15 g/mol
MDL: MFCD00139060
CAS-Nummer: 69155-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR