6,8-Dimethylchromon-2-carbonsäure

Lieferant: CHEMPUR

FL12040.POR 012040-5G
CHMPFL12040.POREA 0 EUR
CHMPFL12040.POR FLUO012040-5G
6,8-Dimethylchromon-2-carbonsäure
6,8-Dimethylchromon-2-carbonsäure
Formel: C₁₂H₁₀O₄
MDL: MFCD03424503
CAS-Nummer: 288399-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR