Dumont AA, epoxidbeschichtete Zange
Lieferant: FINE SCIENCE TOOLS
11210-10
FSCI11210-10EA 42 EUR
FSCI11210-10
Dumont AA, epoxidbeschichtete Zange
Pinzetten
Slip-proof acid and electric shock resistant epoxy coating covers the forceps 17,5 mm from the tips.