Dextran, M.W. 60000 - 90000, niedriger Anteil, J.T.Baker®
Lieferant: Avantor
G200-05EA
238.52
EUR
G200-05
Dextran, M.W. 60000 - 90000, niedriger Anteil, J.T.Baker®
Dextran
Formel:
(C₆H₁₀O₅)n |
MDL:
MFCD00130935 CAS-Nummer: 9004-54-0 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...