AAV1/2-CMV/CBA-Human-A53T-alpha-SYN AAV Vector
Lieferant: Charles River Laboratories Cell Solutions
GD1001-RV
GD1001-RVEA 0 EUR
GD1001-RV
AAV1/2-CMV/CBA-Human-A53T-alpha-SYN AAV Vector
Vektoren, Plasmide und Bibliotheken
AAV1/2 Vector with CMV promoter, human A53T alpha-synuclein, WPRE, bovine growth hormone (BGH), and polyadenylation sequence.
  • CMV promoter
  • AAV1/2 Serotype
  • PBS with 1 mM MgCl₂ and 0,001% Lutrol F68 storage buffer
The CMV/CBA promoter consists of the chicken beta-actin promoter hybridised with the CMV immediate early enhancer sequence and is highly efficient in most tissue types. The Woodchuck post-transcriptional regulatory element (WPRE) and the presence of a bovine growth hormone (BGH) polyadenylation sequence ensures high transcription following transduction.
Shipped in dry ice.