Test tubes, Durham
Lieferant: GERBER FUNKE
8130
GERB8130EA 12.75 EUR
GERB8130
Test tubes, Durham
Röhrchen Reagenzgläser- und Probenröhrchen
Glass, clear.
  • Excellent chemical resistance
  • Good temperature resistance