Hämatoxylin-Lösung Gill II, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DcH-48, Natural Black I, Hydroxybrasilin

M-Clarity™ Qualitätsstufe = MQ500

GHS2128-4L
GHS2128-4LEA 295 EUR
GHS2128-4L
Hämatoxylin-Lösung Gill II, Sigma-Aldrich®
Haematoxylin staining solutions

Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.


  • 4 g/L certified hematoxylin

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00078111
CAS-Nummer: 517-28-2

Bestellen

Spezifikation Testergebnisse

Appearance (Turbidity) Clear
Appearance(Colour)
Reddish-Purple
Conforms
Appearance (Form) Solution
Essentially free of precipitate
Performance Conforms
Using Sigma-Aldrich Procedure No. GHS, Nuclei and compontents stain blue to black, Chromatin clearly visible
Expiration Date Period 24 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR