Hämatoxylin Gill No. 2, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DcH-48, Natural Black I, Hydroxybrasilin

GHS232-1L
GHS232-1LEA 97.4 EUR
GHS232-1L
Hämatoxylin Gill No. 2, Sigma-Aldrich®
Haematoxylin staining solutions

Gill formulation No. 2 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078111
CAS-Nummer: 517-28-2
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR