Hämatoxylin Gill No. 3, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DcH-48, Natural Black I, Hydroxybrasilin

GHS332-1L
GHS332-1LEA 118 EUR
GHS332-1L
Hämatoxylin Gill No. 3, Sigma-Aldrich®
Haematoxylin staining solutions

Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time. Used with hematoxylin and eosin staining.

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078111
CAS-Nummer: 517-28-2
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR