Hämatoxylin-Lösung Gill III, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: DcH-48, Natural Black I, Hydroxybrasilin

M-Clarity™ Qualitätsstufe = MQ500

GHS380-2.5L
GHS380-2.5LEA 225 EUR
GHS380-2.5L
Hämatoxylin-Lösung Gill III, Sigma-Aldrich®
Haematoxylin staining solutions

Gill formulation No. 3 may be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time. Used with hematoxylin and eosin staining.


  • 6 g/L certified hematoxylin

Formel: C₁₆H₁₄O₆
Molecular Weight: 302,28 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00078111
CAS-Nummer: 517-28-2

Bestellen

Spezifikation Testergebnisse

Appearance (Colour)
Dark Reddish-Purple
Conforms
Appearance (Form) Solution
Appearance (Turbidity)
Essentially free of precipitate
Clear
Performance Conforms
Using Sigma-Aldrich Procedure GHS, nuclei and components stain blue to black; chromatin clearly visible.
Expiration Date Period 24 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR