Slide box
Lieferant: GLW
K05
GLWAK05EA 4.7 EUR
GLWAK05
Slide box
Objektträger-Kästen
Slide box with five compartments and fixed blade to secure the slides.
  • Slides are stabilised with the fixed blade in the film hinged lid
  • Can be used in freezer down to –90 °C
  • Optional safety seals can prevent the unauthorised access
Suitable for storing, transporting, freezing and autoclaving the samples.