Straw pipettes, STRAWS 180
Lieferant: Grade Products
STW180
GRADSTW180EA 157 EUR
GRADSTW180
Straw pipettes, STRAWS 180
Pipetten Saugpipetten
These disposable pipette straws are specifically designed for use with laboratory blender bags. They are sterile, unbreakable and eliminate the hazard of broken glass. They also avoid exposure to infectious materials and contaminating samples.
Length: 180 mm
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904