Lubricant, anti-friction coating, Molykote® D-321 R
Lieferant: GREEFF RW SILICONES LUBRICAN
636264F
GREF636264FEA 0 EUR
GREF636264F
Lubricant, anti-friction coating, Molykote® D-321 R
Schmiermittel
Air-curing dry film lubricant in solvent.
  • Air drying
  • Avoidance of stick-slip
  • High aging resistance
Recommended dry film thickness: 5 to 20 µm
Service temperature range from –180 to +260 °C.