Digital vacuum - / barometer, GMH3161-12
Lieferant: Greisinger
600407
GRSG600407EA 0 EUR
GRSG600407
Digital vacuum - / barometer, GMH3161-12
Barometer
This handheld meter with integrated absolute pressure sensor is suitable for measurements in air and non corrosive and non ionizing gases and liquids.
  • Impact resistant ABS, membrane keyboard, transparent panel, front side with IP65 rating
  • Min./Max. memory, HOLD function, automatic power-off
  • Sea level correction
  • Piezo-resistive absolute pressure sensor integrated in device
  • Serial interface (3,5 mm jack) can be connected to RS232 or USB interface of a PC via electrically isolated interface adapter (needs to be ordered separately)
Lieferumfang: Supplied with batteries and manual.