tert-Butyldichlorophosphine 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
t-Butyldichlorophosphine
H27360.18
H27360.18EA
520
EUR
H27360.18
tert-Butyldichlorophosphine 98%
tert-Butyldichlorophosphine
Purity: 98%
CAS No.: 25979-07-1
Molecular Formula: C4H9Cl2P
Formula Weight: 159.00
Melting Point: 44-49°
UN No.: UN3261
MDL No.: MFCD00013615
Formel:
C₄H₉Cl₂P Molecular Weight: 158,99 g/mol Schmelzpunkt: 44…49 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013615 CAS-Nummer: 25979-07-1 EINECS: 247-386-9 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...