2-Methoxybenzenesulphonyl chloride 97%

Lieferant: Thermo Scientific
Gefahr

H34455.03
H34455.03EA 131 EUR
H34455.03
2-Methoxybenzenesulphonyl chloride 97%
2-Methoxybenzenesulphonyl chloride

Purity: 97% CAS No.: 10130-87-7 Molecular Formula: C7H7ClO3S Formula Weight: 206.65 Melting Point: 50-54° UN No.: UN3261 MDL No.: MFCD01961367

Formel: C₇H₇ClO₃S
Molecular Weight: 206,65 g/mol
Schmelzpunkt: 50…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01961367
CAS-Nummer: 10130-87-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR