Filtertrichter, mikro
Lieferant: Hach
2584335
HACH2584335EA 8.2 EUR
HACH2584335
Filtertrichter, mikro
Trichter Filtertrichter Filtertrichter
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
Ideal for micro work, filling burets, manometers, and small bottles.