Dissolved oxygen probe, polarographic, sensION™+ 5130
Lieferant: Hach
LZW5130970002
HACHLZW5130970002EA 685 EUR
HACHLZW5130970002
Dissolved oxygen probe, polarographic, sensION™+ 5130
Elektroden Sauerstoff-Elektroden
Polarographic dissolved oxygen (DO) probe with built-in temperature sensor. For use with portable meters.
  • With 1 meter cable and MP5 connector
  • Durable plastic body
  • Replaceable oxygen-permeable membrane, refillable reference electrolyte
The electrode is ideal for dissolved oxygen measurement in general aqueous applications – particularly in a field environment.
Lieferumfang: Supplied with 30 ml bottle of electrolyte solution, two membrane modules and polishing strips.