Analogue stopwatch
Lieferant: HANHART
121.0117-00
HANH121.0117-00EA 297 EUR
HANH121.0117-00
Analogue stopwatch
Timer Stoppuhren, analog
Addition timer in functional ABS case.
  • Pin lever, 1 jewel
  • Timing capacity: 30 minutes
  • Very lightweight and easy to handle
  • Shock protected