Analogue stopwatch, Allsport
Lieferant: HANHART
122.6401-00
HANH122.6401-00EA 453 EUR
HANH122.6401-00
Analogue stopwatch, Allsport
Timer Stoppuhren, analog
Universal referee’s stopwatch/addition timer in chromium plated brass case.
  • Pin lever, 7 jewels
  • Scale goes up to 60 minutes
  • 20, 35, 45 and 60 minutes marked in yellow
  • Shock-, dust- and waterproof