Cell spreaders, Delta™
Lieferant: HEATHROW
HS8151
HEATHS8151EA 298 EUR
HEATHS8151
Cell spreaders, Delta™
Schaber Zellschaber
Triangular shaped spreading bar has an edgeless design which prevents tearing of the agar and head shape that distributes pressure evenly for smooth spreading. Handle flexes slightly under pressure and is rippled for easy manipulation. Dimensions 172×37×4 mm.