Cell spreaders, L-shaped
Lieferant: HEATHROW
HS8171B
HEATHS8171BEA 171 EUR
HEATHS8171B
Cell spreaders, L-shaped
Schaber Zellschaber
ABS disposable L-shaped cell spreader with smooth surface that will not tear agar plate or disturb media layer. Supplied sterile for immediate use without flaming.