Bath fluids, Aquaclean
Lieferant: GLASWARENFABRIK KARL HECHT
43170003
HECH43170003EA 35.9 EUR
HECH43170003
Bath fluids, Aquaclean
Flüssigkeiten für Umwälzbäder
Additive for keeping water bath and water circulating systems clean.
  • Prevents algae and bacterial growth
  • Prevents odour formation and obstructions in tubing and temperature control system
  • Saving of time: frequent cleaning of the tank is unnecessary
  • Solution is without aggressive phosphates and oxidising or reducing substances
  • Prevents foam formation, even with strong circulation
Bitte beachten Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.