Bath fluids, Aquaclean
Lieferant: GLASWARENFABRIK KARL HECHT
43170003
HECH43170003EA
35.9
EUR
HECH43170003
Bath fluids, Aquaclean
Flüssigkeiten für Umwälzbäder
Additive for keeping water bath and water circulating systems clean.
- Prevents algae and bacterial growth
- Prevents odour formation and obstructions in tubing and temperature control system
- Saving of time: frequent cleaning of the tank is unnecessary
- Solution is without aggressive phosphates and oxidising or reducing substances
- Prevents foam formation, even with strong circulation
Bitte beachten Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.