Cuvettes for fluorescence measurements, QS/QG
Lieferant: HELLMA
FS4C-10103M50L FS4C-10103M50S
HELLFS4C-10103M50LEA 0 EUR
HELLFS4C-10103M50L HELLFS4C-10103M50S
Cuvettes for fluorescence measurements, QS/QG
Küvetten Spektralphotometer-Küvetten
These general-purpose cuvettes are mainly designed for fluorescence measurements in the UV/visible wavelength range.
  • Rectangular form
  • Classified as a macro cell
  • Made of quartz suprasill/quartz glass
Designed with four optical windows for static sampling and hence this cuvette will fit all standard spectrophotometer cell holders and can be used with any beam height.