Spray chamber
Lieferant: Agilent
G1820-65337
HEWLG1820-65337EA 0 EUR
HEWLG1820-65337
Spray chamber
Zerstäuber
This spray chamber made of quartz is used with 7500 ICP-MS.
It is dedicated for babington, concentric and micro-concentric nebulisers.
Bitte beachten Not suitable for hydrofluoric acid.