Spray chamber
Lieferant: Agilent
G3139-65003
HEWLG3139-65003EA 3660 EUR
HEWLG3139-65003
Spray chamber
Zerstäuber
This PFA spray chamber is the standard spray chamber used with the 7500 ICP-MS when aspirating hydrofluoric acid.