Replacement plunger, for blue line autosampler
Lieferant: Agilent
G4513-80227
HEWLG4513-80227EA 0 EUR
HEWLG4513-80227
Replacement plunger, for blue line autosampler
Spritzenzylinder, Stößel und Kolben
PTFE tip plunger for blue line autosampler fixed needle syringes for 7693A ALS.