Avantor® Hichrom, ValvTool
Lieferant: Avantor
HI-199EA 74 EUR
HI-199
Avantor® Hichrom, ValvTool
Schraubenschlüssel
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¼" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.