Embedding system, Technovit® 3040
Lieferant: HERAEUS KULZER
64708805
HKUL64708805EA 168 EUR
HKUL64708805
Embedding system, Technovit® 3040
Gewebeeinbetter
Technovit 3040 is a quickly hardening two-component plastic with a methyl meth acrylate base (MMA).
  • Chemical composition allows for a stable permanent bond with the hardened block
  • Hardens and the Histobloc tightly bonds to the specimen
  • Specimen can now be removed from the form and is ready to be cut. Through blocking with the very hard
Used to block or fasten embedded plastic specimens via a block or emmbedding specimen.