Embedding system, Technovit® 9100
Lieferant: HERAEUS KULZER
64715444
HKUL64715444EA 392 EUR
HKUL64715444
Embedding system, Technovit® 9100
Gewebeeinbetter
Technovit 9100 is a plastic embedding system used in medicine, botany and zoology.
  • Uniform block hardening
  • PMMA block remains transparent
  • Polymerisation below freezing
Designed for mineralised tissues as well as soft tissue with an expanded study spectrum in light microscopy.
The deplasticised sections are suitable for histological overview staining, enzyme chemistry and immunohistological studies, including in situ hybridisation.
Lieferumfang: Contains a basic solution (1000 ml), PMMA powder (120 g), hardners 1 (8×1 g), hardners 2 (1×10 ml) and a regulator (5 ml).