Sleeve protectors, PP
Lieferant: HUBEI ORIENT INTL TRADING
HOIT109760EA 89.8 EUR
HOIT109760
Sleeve protectors, PP
Einweg-Kleidung
These disposable sleeve protectors made of PP provide waterproof protection for light-duty tasks.
  • Lightweight and soft
  • Waterproof and oil-proof
  • Breathable fabric
  • Latex-free
Ideal for use in non-hazardous applications, such as cleaning, maintenance, food processing and handling, manufacturing, hotels and catering.
Weight - 40 g/m²