Flußsäure 49,5-50,5%, Halbleiter-Qualität (Honeywell 17736)

Lieferant: Honeywell Chemicals
Gefahr

40213H-5L
HONC40213H-5LEA 166.69 EUR
HONC40213H-5L
Flußsäure 49,5-50,5%, Halbleiter-Qualität (Honeywell 17736)
Hydrofluoric acid

Beilstein Number: 1098214

Formel: HF
Molecular Weight: 20,01 g/mol
Siedepunkt: 105 °C (1,013 hPa)
Schmelzpunkt: –35 °C
Dichte: 1,170 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay 49.5 - 50.5 %
Residue on ignition (SO₄) Max. 5 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.05 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.05 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.2 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.1 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 5 ppm
Hexafluorosilicic acid (H₂SiF₆) Max. 50 ppm
Nitrate (NO₃) Max. 3 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR