Salpetersäure ≥65%, Halbleiter-Qualität (Honeywell 17818)
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Nitric acid solution
40230H-2.5L
HONC40230H-2.5LEA
146.15
EUR
HONC40230H-2.5L
Salpetersäure ≥65%, Halbleiter-Qualität (Honeywell 17818)
Salpetersäure
RTECS Number: 231-714-2
Formel:
HNO₃ Molecular Weight: 63,01 g/mol Siedepunkt: 122 °C (1,013 hPa) Schmelzpunkt: –31 °C Dichte: 1,400…1.480 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011349 CAS-Nummer: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II |
Spezifikation Testergebnisse
Assay | Min. 65 % |
Residue on ignition (SO₄) | Max. 5 ppm |
Silver (Ag) | Max. 0.01 ppm |
Aluminium (Al) | Max. 0.05 ppm |
Arsenic (As) | Max. 0.01 ppm |
Gold (Au) | Max. 0.02 ppm |
Boron (B) | Max. 0.05 ppm |
Barium (Ba) | Max. 0.01 ppm |
Beryllium (Be) | Max. 0.01 ppm |
Bismuth (Bi) | Max. 0.02 ppm |
Calcium (Ca) | Max. 0.1 ppm |
Cadmium (Cd) | Max. 0.01 ppm |
Cobalt (Co) | Max. 0.01 ppm |
Chromium (Cr) | Max. 0.01 ppm |
Copper (Cu) | Max. 0.01 ppm |
Iron (Fe) | Max. 0.1 ppm |
Gallium (Ga) | Max. 0.02 ppm |
Germanium (Ge) | Max. 0.05 ppm |
Indium (In) | Max. 0.02 ppm |
Potassium (K) | Max. 0.1 ppm |
Lithium (Li) | Max. 0.01 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.01 ppm |
Molybdenum (Mo) | Max. 0.01 ppm |
Sodium (Na) | Max. 0.3 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.01 ppm |
Platinum (Pt) | Max. 0.02 ppm |
Antimony (Sb) | Max. 0.01 ppm |
Tin (Sn) | Max. 0.02 ppm |
Strontium (Sr) | Max. 0.01 ppm |
Titanium (Ti) | Max. 0.01 ppm |
Thallium (Tl) | Max. 0.02 ppm |
Vanadium (V) | Max. 0.01 ppm |
Zinc (Zn) | Max. 0.05 ppm |
Zirconium (Zr) | Max. 0.01 ppm |
Chloride (Cl) | Max. 0.5 ppm |
Phosphate (PO₄) | Max. 0.5 ppm |
Sulfate (SO₄) | Max. 0.5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...