Salzsäure ≥32%, Halbleiter-Qualität (Honeywell 17863)

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydrochloric acid solution, Muriatic acid

40233H-2.5L
HONC40233H-2.5LEA 138.91 EUR
HONC40233H-2.5L
Salzsäure ≥32%, Halbleiter-Qualität (Honeywell 17863)
Salzsäure

Beilstein Number: 1098214

Formel: HCl
Molecular Weight: 36,46 g/mol
Siedepunkt: 42 °C (1,013 hPa)
Schmelzpunkt: –35 °C
Dichte: 1,190 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
UN: 1789
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay Min. 32 %
Residue on ignition (SO₄) Max. 5 ppm
Ammonium (NH₄) Max. 2 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.02 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.02 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.01 ppm
Bromide (Br) Max. 50 ppm
Free chlorine (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR