2-Propanol ≥99,8% VLSI, Halbleiter-Qualität (Honeywell 17604)

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol

40301H-2.5L
HONC40301H-2.5LEA 246.28 EUR
HONC40301H-2.5L
2-Propanol ≥99,8% VLSI, Halbleiter-Qualität (Honeywell 17604)
2-Propanol

Beilstein Number: 635640
RTECS Number: NT8050000
UNSPSC Code: 12191601

Formel: (CH₃)₂CHOH
Molecular Weight: 60,1 g/mol
Siedepunkt: 77-83 °C (1,013 hPa)
Schmelzpunkt: -90 °C
Dichte: 0,784…0.786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011675
CAS-Nummer: 67-63-0
EINECS: 200-661-8
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Particles > 0,5 µm Max. 250 P/ml
Assay (GC) Min. 99.8 %
Boiling range 81 - 83 °C
Density (D 20/4) 0.785 - 0.787
Refractive index (n 20/D) 1.3770 - 1.3780
Non-volatile matter Max. 5 ppm
Water (Karl Fischer) Max. 500 ppm
Free acid (as C₂H₅COOH) Max. 10 ppm
Silver (Ag) Max. 0.01 ppm
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Gold (Au) Max. 0.02 ppm
Boron (B) Max. 0.01 ppm
Barium (Ba) Max. 0.02 ppm
Beryllium (Be) Max. 0.01 ppm
Bismuth (Bi) Max. 0.02 ppm
Calcium (Ca) Max. 0.05 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.01 ppm
Copper (Cu) Max. 0.01 ppm
Iron (Fe) Max. 0.05 ppm
Gallium (Ga) Max. 0.01 ppm
Indium (In) Max. 0.01 ppm
Potassium (K) Max. 0.02 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.02 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.01 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.01 ppm
Lead (Pb) Max. 0.01 ppm
Platinum (Pt) Max. 0.02 ppm
Antimony (Sb) Max. 0.01 ppm
Tin (Sn) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.01 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.02 ppm
Zirconium (Zr) Max. 0.01 ppm
Chloride (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 1 ppm
Acetone (GC) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR